Skip to Content
MilliporeSigma
All Photos(1)

Documents

R5030

Sigma-Aldrich

Ridaifen-B

≥98% (HPLC), white, solid

Synonym(s):

1,1′-[(2-Phenyl-1-buten-1-ylidene)bis(4,1-phenyleneoxy-2,1-ethanediyl)]bis-pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H42N2O2
CAS Number:
Molecular Weight:
510.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

availability

not available in Japan

storage condition

desiccated

color

white

solubility

DMSO: ≥13 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CC\C(c1ccccc1)=C(/c2ccc(OCCN3CCCC3)cc2)c4ccc(OCCN5CCCC5)cc4

InChI

1S/C34H42N2O2/c1-2-33(28-10-4-3-5-11-28)34(29-12-16-31(17-13-29)37-26-24-35-20-6-7-21-35)30-14-18-32(19-15-30)38-27-25-36-22-8-9-23-36/h3-5,10-19H,2,6-9,20-27H2,1H3

InChI key

XQQWCGJGUHJSLR-UHFFFAOYSA-N

Biochem/physiol Actions

Ridaifen-B (RID-B) is a novel tamoxifen (TAM) analog that significantly augments apoptosis-inducing effect of TAM in estrogen receptor (ER)-negatives cells. Ridaifen-B induces mitochondria-involved apoptosis in Jurkat cells, as evidenced by chromatin-condensed cells as well as downstream activation of caspases (caspase-3, -8 and -9) in a dose- and time-dependent manner. At 4 hours of incubation, IC50 for RID-B is 4 muM (30 muM for TAM). And at prolonged treatment of 48 hours, IC50 for RID-B is 0.1 muM. In a related report on the global anti-tumor activity, RID-B strongly inhibits 39 human cancer cells (JFCR 39), both ER-+ or ER-- at concentrations of equal or less than 1muM (e.g., at 0.38muM for SF-539 [central nervous system], at 0.58muM for HT-29 [colon], at 0.20muM for DMS114 [lung], at 0.21muM for LOX-IMVI [melanoma], and at 0.23muM for MKN74 [stomach]. The binding protein of RID-B that exerts the apoptosis events is currently under investigation.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service