Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R4506

Sigma-Aldrich

Msp I from Moraxella sp.

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-C/CGG-3′
Ligation and recutting results: After 2-10-fold Msp I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: Inactivated at 65 °C for 15 minutes.

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SL (B3782).

Physical form

Solution in 10 mM Tris-HCl, pH 7.5, 1 mM EDTA, 50 mM KCl, 10 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.2% Triton X-100 (v/v) at 4 °C

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Waalwijk et al.
Nucleic acids research, 5(9), 3231-3236 (1978-09-01)
The cleavage of DNA by restriction endonucleases HpaII and HapII is prevented by the presence of a 5-methyl group at the internal C residue of its recognition sequence CCGG. MspI, an isoschizomer of HpaII available from New England Biolabs, cleaves
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Nico Mitro et al.
Methods in molecular biology (Clifton, N.J.), 952, 137-144 (2012-10-27)
The role of certain amino acids in the interactions of ligands with their cognate nuclear receptors is usually achieved by the resolution of the crystal structure of the receptor complexed with the ligand. As a complementary functional approach, site-directed mutagenesis
Rachel M Smith et al.
Nucleic acids research, 41(1), 391-404 (2012-11-14)
Type IIB restriction-modification systems, such as BcgI, feature a single protein with both endonuclease and methyltransferase activities. Type IIB nucleases require two recognition sites and cut both strands on both sides of their unmodified sites. BcgI cuts all eight target
Yu-Feng Huang et al.
BMC systems biology, 6 Suppl 2, S10-S10 (2013-01-11)
Current next-generation sequencing (NGS) platforms adopt two types of sequencing mechanisms: by synthesis or by ligation. The former is employed by 454 and Solexa systems, while the latter by SOLiD system. Although the pros and cons for each sequencing mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service