Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R4155

Sigma-Aldrich

Anti-hnRNP-E1/E2 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HNRPE1/HNRPE2, Anti-Heterogeneous nuclear ribonucleoprotein E1/E2, Anti-PCBP1/PCBP2, Anti-poly(rC) binding protein 1 /protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using K562 cell extract
western blot: 0.5-1 μg/mL using K562 cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCBP1(5093)
mouse ... Pcbp1(23983)
rat ... Pcbp1(500242)

General description

hnRNP-E1/E2 (heterogeneous nuclear ribonucleoproteins) belongs to hnRNP (heterogeneous ribonucleoprotein) family. It is also known as poly-C binding proteins (PCBP) 1 and 2. It is located on chromosome 2p13. PCBP1 gene has no intron and codes for a 356 amino acid poly (rC) binding protein 1, that is of 35kDa. It has three K-Homology (KH) domains and two nuclear localization signals.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

hnRNP-E1/E2 (heterogeneous nuclear ribonucleoproteins) participates in the development and progression of tumor. It is involved in several transcriptional and posttranscriptional events like mRNA transcription regulation, transportation, splicing, translation, and stabilization. It plays critical roles in cancer apoptosis, metastasis, invasion and malignant transformation. It may also participates in epithelial-mesenchymal transition (EMT).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulated Poly-C binding protein-1 is a novel predictor associated with poor prognosis in Acute Myeloid Leukemia
Zhou M and Tong X
Diagnostic Pathology, 10:147 (2015)
Molecular determinants of the axonal mRNA transcriptome
Gomes C, et al.
Developmental neurobiology, 74(3), 218-232 (2014)
Expression of poly(C)-binding protein 1 (PCBP1) in NSCLC as a negative regulator of EMT and its clinical value
Liu Y, et al.
International Journal of Clinical and Experimental Pathology, 8(6), 7165-7172 (2015)
The PCBP1 gene encoding poly(rC) binding protein I is recurrently mutated in Burkitt lymphoma
Wagener R, et al.
Genes Chromosomes Cancer, 54(9), 555-564 (2015)
Emerging novel functions of the oxygen-sensing prolyl hydroxylase domain enzymes
Wong BW, et al.
Trends in Biochemical Sciences, 38(1), 3-11 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service