Skip to Content
MilliporeSigma
All Photos(2)

Documents

R306-05A

Sigma-Aldrich

Rat Cardiac Fibroblasts: RCF, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat heart (Sprague Dawley, normal adult)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 42

morphology

fibroblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Cardiac fibroblasts are the most prevalent cell type in the heart, making up 60-70 % of all cells. They are known to play key roles in the development of cardiac disease and hypertrophy. Rat Cardiac Fibroblasts provide an excellent model system to study many aspects of heart function and pathophysiology, including the regulation of cell growth, survival, inflammation, hormone secretion and metabolism.

Cell Line Origin

Heart

Application

human heart function, neointimal formation, vascular remodeling, myocardial repair, extracellular maintenance, signaling, toxicology, gene expression, structural support for cardiomycytes, synthesis of extracellular matrix, growth factors and cytokines

Components

Fibroblast Basal Medium that contains 10% FBS and 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Rat Fibroblast Basal Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 8 population doublings

Subculture Routine

Please refer to the RCF Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service