Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2654

Sigma-Aldrich

Anti-p190 RhoGAP (GK-18) antibody produced in rabbit

~0.8 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GRLF1, Anti-Glucorticoid Receptor Binding Factor 1, Anti-p190

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 190 kDa

species reactivity

canine, human, mouse

concentration

~0.8 mg/mL

technique(s)

immunoprecipitation (IP): 20-40 μg using MDCK cell lysate
western blot: 1-2 μg/mL using mouse brain extract (S1 fraction), MDCK HeLa whole cell extract.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRLF1(2909)
mouse ... Grlf1(232906)
rat ... Grlf1(306400)

General description

The p190 RhoGAP protein is also termed as p190A or glucocorticoid receptor binding factor 1 (GRLF1) 190 kDa. It has three major regions, such as an N-terminal GTP binding domain, a middle region and a C-terminal GTPase-activating proteins (GAP) domain. Rho GTPase activating protein 35 (ARHGAP35) codes for p190A RhoGAP (p190A) protein. It is an important GTPase-activating protein (GAP), which belongs to the Rho GTPase-activating proteins (RhoGAP) family. This gene is located on human chromosome 19q13.3

Specificity

Anti-p190 RhoGAP (GK-18) specifically recognizes p190 RhoGAP (190 kDa).

Immunogen

synthetic peptide corresponding to amino acids 1131-1148 located near the C-terminus of rat p190 RhoGAP, conjugated to KLH.

Application

Anti-p190 RhoGAP (GK-18) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

The p190 RhoGAP protein or glucorticoid receptor binding factor 1 (GRLF1) serves as a GTPase-activating protein (GAP) for the Rho and Rac family of proteins, that are involved in regulating the reorganization of cytoskeletal actin and membrane ruffling. It controls axonal growth and guidance. p190 RhoGAP is essential for normal neural development. p190 RhoGAP has been shown to be tyrosine phosphorylated by c-Src and v-Src both in vitro and in vivo. The middle domain region of this protein mediates binding to Ras GTPase activating protein (p120 RasGAP). p190A plays a key role in the development of mouse and tissue homeostasis. In mammals, p190A also participates in the proliferative and motile capacities of cells. Mutations in the gene is implicated in cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p190 RhoGAP promotes contact inhibition in epithelial cells by repressing YAP activity
Frank SR, et al.
The Journal of cell biology, 217(9), 3183-3201 (2018)
The adhesion signaling molecule p190 RhoGAP is required for morphogenetic processes in neural development
Brouns MR, et al.
Development, 127(22), 4891-4903 (2000)
Molecular cloning, characterization and mapping of the canine glucocorticoid receptor DNA binding factor 1 (GRLF1)
Zangerl B, et al.
Gene, 294(1-2), 167-176 (2002)
p190 RhoGAP is the principal Src substrate in brain and regulates axon outgrowth, guidance and fasciculation
Brouns MR, et al.
Nature Cell Biology, 3(4), 361-367 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service