Skip to Content
MilliporeSigma
All Photos(1)

Documents

R1780

Sigma-Aldrich

RPMI-1640 Medium

With ʟ-glutamine, sodium bicarbonate without arginine, leucine, lysine, and phenol red, sterile liquid, suitable for cell culture, suitable for isotope labeling for cell culture applications

Synonym(s):

RPMI 1640 Cell Culture Medium, Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: 2 g/L
phenol red: no
glucose: 2 g/L (Dextro)
L-glutamine: 0.3 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Reconstitution

Designed for use in stable isotope labeling with amino acids in cell culture applications.
Supplement with 0.100 g/L L-Arginine-13C6, 15N4 HCl (Product 608033), 0.100 g/L L-Leucine-13C6, 15N (Product 608068), and 0.100 g/L L-Lysine-13C6, 15N2 (Product 608041).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengying Ke et al.
Frontiers in oncology, 11, 779562-779562 (2021-11-23)
Multiple myeloma (MM) is a hematological malignancy worldwide in urgent need for novel therapeutic strategies. Since Velcade (bortezomib) was approved for the treatment of relapsed/refractory MM in 2003, we have seen considerable improvement in extending MM patient survival. However, most
Hong-Ren Yu et al.
Nutrients, 13(8) (2021-08-28)
l-Arginine is an important nutrient in the infant diet that significantly regulates the maturation of the immune system in neonates, including the maturation of CD4+ T cells. The biological activities of CD4+ T cells differ substantially between neonates and adults
Rachel S Lee et al.
Neoplasia (New York, N.Y.), 21(4), 389-400 (2019-03-23)
Approximately 50% of prostate cancers harbor the TMPRSS2:ERG fusion, resulting in elevated expression of the ERG transcription factor. Despite the identification of this subclass of prostate cancers, no personalized therapeutic strategies have achieved clinical implementation. Kinases are attractive therapeutic targets
Zhang Xiao Ying et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(21), 5383-5393 (2016-11-03)
This study was performed to further our understanding of the biological and genetic basis of follicular lymphoma and to identify potential novel therapy targets. We analyzed previously generated whole exome sequencing data of 23 follicular lymphoma cases and one transformed
Adele Goldman-Pinkovich et al.
mBio, 11(5) (2020-10-15)
Arginine homeostasis in lysosomes is critical for the growth and metabolism of mammalian cells. Phagolysosomes of macrophages are the niche where the parasitic protozoan Leishmania resides and causes human leishmaniasis. During infection, parasites encounter arginine deprivation, which is monitored by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service