Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R1658

Sigma-Aldrich

RS-102221 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

4-dione hydrochloride hydrate, 8-[5-(2,4-Dimethoxy-5-(4-trifluoromethylphenylsulphonam ido)phenyl-5-oxopentyl]-1,3,8-triazaspiro[4.5]decane-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31F3N4O7S·HCl · xH2O
CAS Number:
Molecular Weight:
649.08 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.COc1cc(OC)c(cc1NS(=O)(=O)c2ccc(cc2)C(F)(F)F)C(=O)CCCCN3CCC4(CC3)NC(=O)NC4=O

InChI

1S/C27H31F3N4O7S.ClH.H2O/c1-40-22-16-23(41-2)20(33-42(38,39)18-8-6-17(7-9-18)27(28,29)30)15-19(22)21(35)5-3-4-12-34-13-10-26(11-14-34)24(36)31-25(37)32-26;;/h6-9,15-16,33H,3-5,10-14H2,1-2H3,(H2,31,32,36,37);1H;1H2

InChI key

IGACJTZTAQBWKN-UHFFFAOYSA-N

Biochem/physiol Actions

RS-102221 is a selective, high affinity 5-HT2C receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris M Panzini et al.
Journal of neurochemistry, 143(6), 708-721 (2017-09-28)
In humans a chromosomal hemideletion of the 16p11.2 region results in variable neurodevelopmental deficits including developmental delay, intellectual disability, and features of autism spectrum disorder (ASD). Serotonin is implicated in ASD but its role remains enigmatic. In this study we
Ao Li et al.
CNS neuroscience & therapeutics, 27(8), 941-950 (2021-05-12)
General anesthesia has been widely applied in surgical or nonsurgical medical procedures, but the mechanism behind remains elusive. Because of shared neural circuits of sleep and anesthesia, whether serotonergic system, which is highly implicated in modulation of sleep and wakefulness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service