Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

R134

Sigma-Aldrich

Rilmenidine hemifumarate salt

solid

Synonym(s):

N-(Dicyclopropylmethyl)-4,5-dihydro-2-oxazolamine hemifumarate salt, Oxaminozoline hemifumarate salt, Rilmenidene hemifumarate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$738.00

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$738.00

About This Item

Empirical Formula (Hill Notation):
C10H16N2O · 0.5C4H4O4
CAS Number:
Molecular Weight:
238.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

color

white

solubility

DMSO: >10 mg/mL
H2O: insoluble

originator

Servier

SMILES string

OC(=O)\C=C\C(O)=O.C1CN=C(NC(C2CC2)C3CC3)O1.C4CN=C(NC(C5CC5)C6CC6)O4

InChI

1S/2C10H16N2O.C4H4O4/c2*1-2-7(1)9(8-3-4-8)12-10-11-5-6-13-10;5-3(6)1-2-4(7)8/h2*7-9H,1-6H2,(H,11,12);1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

LZFATBMLSYHRTC-WXXKFALUSA-N

Gene Information

human ... NISCH(11188)

Related Categories

Biochem/physiol Actions

Selective I1 imidazoline receptor agonist.

Features and Benefits

This compound is featured on the Imidazoline Binding Sites page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Servier. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Takada et al.
British journal of pharmacology, 120(8), 1575-1581 (1997-04-01)
1. To elucidate the possible involvement of pertussis toxin (PTX)-sensitive G proteins in the post receptor mechanism of alpha 2-adrenoceptors and imidazoline receptors, we examined the effect of pretreatment of the central nervous system with PTX on the antidysrhythmic effect
M A Ozog et al.
Journal of neurochemistry, 71(4), 1429-1435 (1998-09-29)
Rilmenidine, a ligand for imidazoline and alpha2-adrenergic receptors, is neuroprotective following focal cerebral ischemia. We investigated the effects of rilmenidine on cytosolic free Ca2+ concentration ([Ca2+]i) in rat astrocytes. Rilmenidine caused concentration-dependent elevation of [Ca2+]i, consisting of a transient increase
C K Chan et al.
The Journal of pharmacology and experimental therapeutics, 276(2), 411-420 (1996-02-01)
The present study in conscious rabbits with intracisternal (i.c.) catheters sought to determine the relative contribution of the I1 subtype of imidazoline receptors (IR) and alpha 2 adrenoceptors to the hypotensive effects of rilmenidine, clonidine and moxonidine with an I1-IR/alpha
Ju-Hyun Lee et al.
Journal of molecular biology, 432(8), 2633-2650 (2020-02-28)
Lysosomal dysfunction is considered pathogenic in Alzheimer disease (AD). Loss of presenilin-1 (PSEN1) function causing AD impedes acidification via defective vacuolar ATPase (vATPase) V0a1 subunit delivery to lysosomes. We report that isoproterenol (ISO) and related β2-adrenergic agonists reacidify lysosomes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service