Skip to Content
MilliporeSigma
All Photos(1)

Documents

Q110

Sigma-Aldrich

Quinelorane dihydrochloride

crystalline

Synonym(s):

(5aR-trans)-5,5a,6,7,8,9,9a,10-Octahydro-6-propylpyrido[2,3-g]quinazolin-2-amine dihydrochloride, LY-163,502

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N4 · 2HCl
CAS Number:
Molecular Weight:
319.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystalline

color

white

solubility

H2O: >10 mg/mL

SMILES string

Cl.Cl.[H][C@]12CCCN(CCC)[C@]1([H])Cc3cnc(N)nc3C2

InChI

1S/C14H22N4.2ClH/c1-2-5-18-6-3-4-10-7-12-11(8-13(10)18)9-16-14(15)17-12;;/h9-10,13H,2-8H2,1H3,(H2,15,16,17);2*1H/t10-,13-;;/m1../s1

InChI key

WDEMLQIGYYLRRX-OWVUFADGSA-N

Gene Information

human ... DRD2(1813)

Biochem/physiol Actions

D2 dopamine receptor agonist; 2-aminopyridine analog of quinpirole.

Legal Information

Manufactured and sold with the permission of Eli Lilly and Company.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Eaton et al.
The Journal of pharmacology and experimental therapeutics, 268(2), 645-652 (1994-02-01)
The purpose of the present study was to characterize dopaminergic D2 receptor-mediated regulation of catecholaminergic neurons in the diencephalon by examining the acute effects of the potent and selective D2 agonist quinelorane (LY163502; trans-(-)-5,5a,6,7,8,9,9a,10-octahydro-6-propylpyrimido [4,5-g] quinolin-2-amine dihydrochloride dihydrate) on concentrations
M A Charles et al.
Pharmacology, biochemistry, and behavior, 43(4), 1087-1092 (1992-12-01)
LY163502, a selective D2 receptor agonist, has been reported to stimulate sexual behavior in both copulating and noncopulating male rats. Three experiments were conducted to further characterize the role of dopamine on male sexual behavior. In the first experiment, quinelorane
J H Carlson et al.
Synapse (New York, N.Y.), 11(3), 229-248 (1992-07-01)
The rates and patterns of discharge activity exhibited by 16 spontaneously active substantia nigra pars compacta dopamine neurons were studied in halothane-anesthetized rats using three types of quantitative measures: 1) mean discharge rates, 2) population characteristics of interspike interval samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service