Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0400

Sigma-Aldrich

PF-915275

≥98% (HPLC)

Synonym(s):

N-(6-Amino-2-pyridinyl)-4′-cyano[1,1′-biphenyl]-4-sulfonamide; N-(6-Aminopyridin-2-yl)-4′-cyanobiphenyl-4-sulfonamide, PF 00915275, PF 915275, PF-00915275, PF00915275, PF915275

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N4O2S
CAS Number:
Molecular Weight:
350.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

[S](=O)(=O)(Nc3nc(ccc3)N)c1ccc(cc1)c2ccc(cc2)C#N

InChI

1S/C18H14N4O2S/c19-12-13-4-6-14(7-5-13)15-8-10-16(11-9-15)25(23,24)22-18-3-1-2-17(20)21-18/h1-11H,(H3,20,21,22)

InChI key

ZESFDAKNYJQYKO-UHFFFAOYSA-N

Biochem/physiol Actions

PF-915275 is an orally active, potent and selective 11beta-hydroxysteroid dehydrogenase type 1 (11ß-HSD1, HSD11B1) inhibitor (human 11ß-HSD1 Ki <1 nM; cellular IC50 = 5 nM using human 11ß-HSD1-transfected HEK293) with reduced potency toward non-human species (human/monkey/dog hepatocytes EC50 = 20/100/120 nM; mouse 11ß-HSD1 Ki = 750 nM, rat hepatoma EC50 = 14 μM). PF-915275 (0.1-3-mg/kg via nasogastric intubations) inhibits prednisone-to-prednisolone conversion in cynomolgus monkeys (by 87% with 3 mg/kg) in vivo and reduces plasma insulin increase following prednisone & resumed feeding (4 hrs post PF-915275) among overnight fasted monkeys.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling-Ling Chang et al.
BMC pharmacology & toxicology, 19(1), 45-45 (2018-07-20)
Nonylphenol (NP) is an environmental endocrine-disrupting chemical (EDC) detected in human cord blood and milk. NP exposure in developmental periods results in hyperadrenalism and increasing 11β-hydroxysteroid dehydrogenase I (11β-HSD1) activity in an adult rat model. Alleviating 11β-HSD1 activity is therefore
B Ganesh Bhat et al.
The Journal of pharmacology and experimental therapeutics, 324(1), 299-305 (2007-10-09)
Glucocorticoids, through activation of the glucocorticoid receptor (GR), regulate hepatic gluconeogenesis. Elevated hepatic expression and activity of 11beta-hydroxysteroid dehydrogenase type 1 (11betaHSD1) play a key role in ligand-induced activation of the GR through the production of cortisol. Evidence from genetically
Jessica K Hartman et al.
Toxicology and applied pharmacology, 355, 112-126 (2018-05-22)
Rising obesity rates worldwide have socio-economic ramifications. While genetics, diet, and lack of exercise are major contributors to obesity, environmental factors may enhance susceptibility through disruption of hormone homeostasis and metabolic processes. The obesogen hypothesis contends that chemical exposure early
Chunpeng Zou et al.
Oncotarget, 8(56), 96263-96275 (2017-12-10)
11β-HSD1 has been recognized as a potential therapeutic target for type 2 diabetes. Recent studies have shown that hyperglycemia leads to activation of 11β-HSD1, increasing the intracellular glucocorticoid levels. Excess glucocorticoids may lead to the clinical manifestations of cardiac injury.
Ling-Ling Chang et al.
Environmental toxicology and pharmacology, 44, 1-12 (2016-04-10)
We previously observed that nonylphenol (NP) exposure during development resulted in increases in body weight and hyperadrenalism in adult male offspring. The mechanism of hyperadrenalism includes the primary activation of the adrenal gland and the conversion of inactive glucocorticoids to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service