Skip to Content
MilliporeSigma
All Photos(1)

Documents

PZ0341

Sigma-Aldrich

PF-06733804

≥98% (HPLC)

Synonym(s):

6-Amino-5-((1S)-2,2-difluoro-4-(2-(4-(trifluoromethoxy)phenyl)acetyl)cyclopentyloxy)-N-methylnicotinamide, 6-Amino-5-[[(3S)-4,4-difluoro-1-[2-[4-(trifluoromethoxy)phenyl]acetyl]-3-pyrrolidinyl]oxy]-N-methyl-3-pyridinecarboxamide, PF06733804

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19F5N4O4
CAS Number:
Molecular Weight:
474.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C(O[C@H]2CN(C(CC3=CC=C(OC(F)(F)F)C=C3)=O)CC2(F)F)C=C(C(NC)=O)C=N1

Biochem/physiol Actions

PF-06733804 is pan tropomyosin-related kinase (Trk) inhibitor.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service