Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0340

Sigma-Aldrich

PF-06737007

≥98% (HPLC)

Synonym(s):

2-((3R,4S)-3-Fluoro-1-(2-(4-(trifluoromethoxy)phenyl)acetyl)piperidin-4-yloxy)-4-(2-hydroxy-2-methylpropoxy)benzamide, 2-[[(3R,4S)-3-Fluoro-1-[2-[4-(trifluoromethoxy)phenyl]acetyl]piperidin-4-yl]oxy]-4-(2-hydroxy-2-methylpropoxy)benzamide, PF06737007

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$133.00
25 MG
$383.00

$133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$133.00
25 MG
$383.00

About This Item

Empirical Formula (Hill Notation):
C25H28F4N2O6
CAS Number:
Molecular Weight:
528.49
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$133.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

F[C@@H]1CN(C(CC2=CC=C(OC(F)(F)F)C=C2)=O)CC[C@@H]1OC3=C(C(N)=O)C=CC(OCC(C)(C)O)=C3

InChI key

AUTGNDRENPBZKA-UXHICEINSA-N

Biochem/physiol Actions

PF-06737007 is pan tropomyosin-related kinase (Trk) inhibitor.
PF-06737007 is pan tropomyosin-related kinase inhibitor.

Other Notes

This compound was developed by Pfizer for Kinase Phosphatase Biology research. To learn more about Sigma′s partnership with Pfizer and view other authentic, high-quality Pfizer compounds, visit sigma.com/bsm-pfizer.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service