Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0323

Sigma-Aldrich

PF-06767832

≥98% (HPLC)

Synonym(s):

1,5-Anhydro-2,3-dideoxy-3-[[[5-methyl-4-[[4-(4-thiazolyl)phenyl]methyl]-2-pyridinyl]carbonyl]amino]-L-threo-pentitol, N-((3R,4S)-3-Hydroxytetrahydro-2H-pyran-4-yl)-5-methyl-4-[4-(1,3-thiazol-4-yl)benzyl]pyridine-2-carboxamide, PF 6767832

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N3O3S
CAS Number:
Molecular Weight:
409.50
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CN=C(C(N[C@@H]2[C@@H](O)COCC2)=O)C=C1CC3=CC=C(C4=CSC=N4)C=C3

InChI

1S/C22H23N3O3S/c1-14-10-23-19(22(27)25-18-6-7-28-11-21(18)26)9-17(14)8-15-2-4-16(5-3-15)20-12-29-13-24-20/h2-5,9-10,12-13,18,21,26H,6-8,11H2,1H3,(H,25,27)/t18-,21-/m0/s1

InChI key

VVZZHFMLRHJXTO-RXVVDRJESA-N

Biochem/physiol Actions

PF-06767832 is a cell permeable, potent and selective M1 muscarinic acetylcholine receptor Positive Allosteric Modulator (PAM). Studies have shown that PF-06767832 potentiates the activity of acetylcholine (Ach) in vitro.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service