Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0295

Sigma-Aldrich

Tiplaxtinin

≥98% (HPLC)

Synonym(s):

(1-Benzyl-5-(4-(trifluoromethoxy)phenyl)-1H-indol-3-yl)oxoacetic acid; a-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid, PAI 039, PAI-039, Tiplasinin, WAY-168039

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$570.00

$143.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$143.00
25 MG
$570.00

About This Item

Empirical Formula (Hill Notation):
C24H16F3NO4
CAS Number:
Molecular Weight:
439.38
MDL number:
UNSPSC Code:
41106609
PubChem Substance ID:
NACRES:
NA.77

$143.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(C(O)=O)C1=CN(CC2=CC=CC=C2)C3=CC=C(C4=CC=C(OC(F)(F)F)C=C4)C=C31

InChI

1S/C24H16F3NO4/c25-24(26,27)32-18-9-6-16(7-10-18)17-8-11-21-19(12-17)20(22(29)23(30)31)14-28(21)13-15-4-2-1-3-5-15/h1-12,14H,13H2,(H,30,31)

InChI key

ODXQFEWQSHNQNI-UHFFFAOYSA-N

Biochem/physiol Actions

Tiplaxtinin has high oral bioavailability. It is metabolically stable and shows large safety multiples in animal toxicology studies. Tiplaxtinin can be easily synthesized in bulk quantities.[1] This drug also reduces diet-induced obesity in mice.[2]
Tiplaxtinin is a potent and selective PAI-1 inhibitor. Tiplaxtinin demonstrated efficacy in vivo in multiple models of acute arterial thrombosis and has been shown to reduce physiologic PAI-1 activity.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiplaxtinin impairs nutritionally induced obesity in mice.
Lijnen H R, et al.
Thrombosis and Haemostasis, 95(06), 731-737 (2006)
Tiplaxtinin, a novel, orally efficacious inhibitor of plasminogen activator inhibitor-1: design, synthesis, and preclinical characterization.
Elokdah H, et al.
Journal of Medicinal Chemistry, 47(14), 3491-3494 (2004)
Camille Cohen et al.
EMBO molecular medicine, 13(11), e14146-e14146 (2021-11-03)
The mechanisms underlying the development of glomerular lesions during aging are largely unknown. It has been suggested that senescence might play a role, but the pathophysiological link between senescence and lesion development remains unexplained. Here, we uncovered an unexpected role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service