Skip to Content
MilliporeSigma
All Photos(1)

Documents

PZ0246

Sigma-Aldrich

PF-5006739

≥98% (HPLC)

Synonym(s):

4-[4-(4-Fluorophenyl)-1-[1-(3-isoxazolylmethyl)-4-piperidinyl]-1H-imidazol-5-yl]-2-Pyrimidinamine, 4-{4-(4-Fluorophenyl)-1-[1-(1,2-oxazol-3-ylmethyl)piperidin-4-yl]-1H-imidazol-5-yl}pyrimidin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22FN7O
CAS Number:
Molecular Weight:
419.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

FC(C=C1)=CC=C1C2=C(C3=CC=NC(N)=N3)N(C4CCN(CC5=NOC=C5)CC4)C=N2

Biochem/physiol Actions

PF-5006739 is a potent inhibitor of casein kinases 1 delta (CK1δ) and 1 epsilon (CK1ε), key regulators of the suprachiasmatic nucleus (SCN) central clock and also linked to the development of drug addiction through PERIOD proteins (PER1–3) and dopamine- and cAMP-regulated neuronal phosphoprotein (DARPP-32) modulation. PF-5006739 has low nM in vitro potency for CK1δ/ε (IC50 values of 3.9 nM for CK1δ and 17.0 nM for CK1ε) and high kinome selectivity. PF-5006739 induced profound phase delays in circadian periodicity in both nocturnal and diurnal animal models and attenuated opioid drug-seeking behavior in a rodent operant reinstatement model.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aida Villarroel et al.
Cellular and molecular life sciences : CMLS, 77(5), 919-935 (2019-07-18)
Wnt ligands signal through canonical or non-canonical signaling pathways. Although both routes share common elements, such as the Fz2 receptor, they differ in the co-receptor and in many of the final responses; for instance, whereas canonical Wnts increase β-catenin stability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service