Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0219

Sigma-Aldrich

PF-06297470

≥98% (HPLC)

Synonym(s):

1-Methyl-3-(4-methyl-3-pyridinyl)-6-(2-pyridinylmethoxy)-1H-pyrazolo[3,4-b]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N6O
CAS Number:
Molecular Weight:
332.36
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(N=C1C2=C(C)C=CN=C2)C3=C1N=CC(OCC4=NC=CC=C4)=N3

InChI

1S/C18H16N6O/c1-12-6-8-19-9-14(12)16-17-18(24(2)23-16)22-15(10-21-17)25-11-13-5-3-4-7-20-13/h3-10H,11H2,1-2H3

InChI key

RDLSKDOSNKLEAQ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06297470 is a potent negative allosteric modulator of the Metabotropic Glutamate Receptor 5 (mGluR5). PF-06297470 binds to mGluR5 with a Ki of 0.9 nM. The compound PF-06297470 displays slight antagonism of mGluR1 (IC50 = 30.5 mM), with no agonist or antagonist activities against any other mGlu receptor subtype at concentrations up to 50 mM. PF-06297470 reduces dyskinesia in a rodent (MPTP)-rendered Parkinsonian model of L-DOPA-induced dyskinesia (PD-LID).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gopinath S Palanisamy et al.
Toxicologic pathology, 43(7), 995-1003 (2015-06-11)
Three orally administered metabotropic glutamate receptor 5 (mGluR5) negative allosteric modulators caused skin lesions consistent with delayed type-IV hypersensitivity in cynomolgus macaques in 2- and 12-week toxicity studies. Several monkeys developed macroscopic skin lesions in multiple locations after 8 to

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service