Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0206

Sigma-Aldrich

PF-04455242 hydrochloride

≥98% (HPLC)

Synonym(s):

2-Methyl-N-((2′-(pyrrolidin-1-ylsulfonyl)biphenyl-4-yl)methyl)propan-1-amine hydrochloride, N-(2-Methylpropyl)-2′-(1-pyrrolidinylsulfonyl)-[1,1′-biphenyl]-4-methanamine hydrochloride, PF-4455242 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28N2O2S · HCl
CAS Number:
Molecular Weight:
408.99
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution, warmed)

storage temp.

room temp

SMILES string

Cl.CC(C)CNCc1ccc(cc1)-c2ccccc2S(=O)(=O)N3CCCC3

InChI

1S/C21H28N2O2S.ClH/c1-17(2)15-22-16-18-9-11-19(12-10-18)20-7-3-4-8-21(20)26(24,25)23-13-5-6-14-23;/h3-4,7-12,17,22H,5-6,13-16H2,1-2H3;1H

InChI key

WRNZYMZEVIVCFL-UHFFFAOYSA-N

Biochem/physiol Actions

PF-4455242 is a selective k opioid receptor (KOR) antagonist with 20-fold selectivity for kappa over mu receptors (k Ki = 3 nM, μ Ki =64 nM). PF-4455242 is orally active, has good brain penetration in rats and was able to block the effects of the kappa agonist U50488. Studies indicate that k blockade has afforded antidepressant activity in several animal models. PF-4455242 has been in phase I clinical trials for trial for bipolar depression.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to opioid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service