Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0120

Sigma-Aldrich

CP-10447

≥98% (HPLC)

Synonym(s):

3-(4-Bromo-2-methylphenyl)-2-methyl-4(3H)-quinazolinone, 4-Bromomethaqualone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13BrN2O
CAS Number:
Molecular Weight:
329.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I

color

white

solubility

DMSO: ≥10 mg/mL

SMILES string

Cc1cc(Br)ccc1N2C(C)=Nc3ccccc3C2=O

InChI

1S/C16H13BrN2O/c1-10-9-12(17)7-8-15(10)19-11(2)18-14-6-4-3-5-13(14)16(19)20/h3-9H,1-2H3

InChI key

CIYGULLCDQXZNK-UHFFFAOYSA-N

Biochem/physiol Actions

CP-10447 is an inhibitor of microsomal triglyceride transfer protein (MTP, MTTP).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service