Skip to Content
MilliporeSigma
All Photos(3)

Documents

PRS2125

Sigma-Aldrich

Anti-MYD88 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Myeloid differentiation primary response gene (88)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 35 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYD88(4615)

General description

Myeloid differentiation primary response 88 (MYD88) is a cytosolic soluble adaptor protein. It possesses a toll-interleukin 1 receptor domain and a death domain. The gene encoding this protein is localized on human chromosome 3p22.2.

Immunogen

a peptide corresponding to amino acids 233-248 of human MyD88, which differ from mouse by two amino acids.

Biochem/physiol Actions

Myeloid differentiation primary response 88 (MYD88) activates innate immune response to Mycobacterium tuberculosis. It is an activator of nuclear factor κ B (NF-κB).The protein mediates the signaling pathways of toll-like and interleukin-1 receptors. Mutation in the gene encoding MYD88 is associated with diffuse large B-cell lymphoma.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2125.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regional copy number?independent deregulation of transcription in cancer
Nicolas Stransky
Nature Genetics, 38(12) (2006)
Conformational dynamics of cancer-associated MyD88-TIR domain mutant L252P (L265P) allosterically tilts the landscape toward homo-dimerization.
Zhan C
Protein engineering, design & selection : PEDS, 29(9), 347-354 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service