Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0296

Sigma-Aldrich

Goat anti-FUS Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

75 kDa DNA-pairing protein, ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS, amyotrophic lateral sclerosis 6, derived from t(12;16) malignant liposarcoma, fus-like protein, fused in sarcoma, fusion, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, heterogeneous nuclear ribonucleoprotein P2, hnRNP-P2, oncogene FUS, oncogene TLS, translocated in liposarcoma, translocated in liposarcoma protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

P35637

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... FUS(2521)

Immunogen

The epitope recognized by PLA0296 maps to a region between residue 1 and 50 of human Fusion (involved in t(12;16) in malignant liposarcoma) using the numbering given in entry P35637 (GeneID 2521).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

FUS has been identified as a frequent translocation fusion partner of various transcription factors. FUS fusion genes have been shown to be associated with multiple tumor types which include liposarcoma, leukemia, histocytoma, and sarcoma. FUS is a multifunctional RNA-binding protein that associates with the nuclear matrix and Cajal bodies and appears to play a role in splicesome assembly, pre-mRNA splicing, DNA repair, transcriptional regulation and homologous recombination.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service