Skip to Content
MilliporeSigma
All Photos(2)

Documents

PLA0236

Sigma-Aldrich

Rabbit anti-ERK2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ERK, ERK-2, ERK2, ERT1, MAP kinase 1, MAP kinase 2, MAP kinase isoform p42, MAPK 1, MAPK 2, MAPK2, P42MAPK, PRKM1, PRKM2, extracellular signal-regulated kinase 2, mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, protein tyrosine kinase ERK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000
western blot: 1:2,000- 1:10,000

accession no.

NP_002736.3

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... ERK2(5594)

Immunogen

The epitope recognized by PLA0236 maps to a region between residue 310 and 360 of human extracellular signal-regulated kinase-2 (mitogen-activated protein kinase 1) using the numbering given in entry NP_002736.3 (GeneID 5594).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Extracellular signal-regulated kinase 2 (ERK2/p42) is also known as mitogen-activated protein kinase 1 (MAPK1). ERK2 is part of a signal transduction pathway that begins with the small GTPase Ras and continues through a cascade of protein kinases that include Raf and MEK. Activation of this signaling pathway influences many cellular processes such as proliferation, differentiation and survival. ERK2 is very similar in sequence to ERK1/p44 and the activities of both ERK1 and ERK2 are usually considered together as total ERK activity. More recent evidence has shown that ERK1 and ERK2 are not functionally redundant and may have very different and specific roles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service