Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0147

Sigma-Aldrich

Rabbit anti-TIF1 Alpha/TRIM24 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

E3 ubiquitin-protein ligase TRIM24, PTC6, RING finger protein 82, RNF82, TF1A, TIF1, TIF1-alpha, TIF1A, TIF1ALPHA, Tripartite motif-containing protein 24, hTIF1, transcriptional intermediary factor 1, tripartite motif-containing 24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

ChIP: 4-30 μg
immunohistochemistry: 1:200- 1:1,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_056989.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TRIM24(8805)

Immunogen

The epitope recognized by PLA0147 maps to a region between residue 700 and 750 of human Transcriptional Intermediary Factor 1 Alpha (Tripartite Motif-Containing 24 ) using the numbering given in entry NP_056989.2 (GeneID 8805).

Physical form

Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide

Other Notes

TIF1 Alpha/TRIM24 is a member of a family of proteins that control transcription and chromatin remodeling through their interaction with transcription factors. The family of TIF1s (transcriptional intermediary factor 1) includes TIF1-alpha, TIF1-beta, and TIF1-gamma. TIF1-alpha plays an important role in development and mediates transcriptional control by interacting with the AF2 region of nuclear receptors such as estrogen, retinoic acid, and vitamin D3 receptors. TIF1-alpha has been shown to be a repressor of transcription. This transcriptional repression involves histone deacetylation and interaction with members of the heterochromatin protein 1 family.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service