Skip to Content
MilliporeSigma
All Photos(1)

Documents

PLA0115

Sigma-Aldrich

Rabbit anti-DNA-PKcs Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DNA-PK catalytic subunit, DNA-PKcs, DNAPK, DNPK1, HYRC, HYRC1, IMD26, XRCC7, complementing 1, hyper-radiosensitivity of murine scid mutation, p350, p460

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:1,000- 1:10,000

accession no.

NP_008835.5

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... DNA-PKcs(5591)

Immunogen

The epitope recognized by PLA0115 maps to a region between residues 2050 and 2100 of human DNA-Dependent Protein Kinase, catalytic subunit using the numbering given in entry NP_008835.5 (GeneID 5591).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

DNA-PKcs is the catalytic subunit of a DNA-dependent serine-threonine protein kinase (DNA-PK). DNA-PK is a heterotrimer comprised of DNA-PKcs and the Ku709/Ku86 dimer. DNA-PK is involved in DNA damage repair and V(D)J recombination. DNA-PK phosphorylates Artemis, a protein required for nonhomologous end-joining and is necessary for the loading of Artemis on damaged DNA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service