Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0066

Sigma-Aldrich

Goat anti-BRD2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

D6S113E, FSH, FSRG1, KIAA9001, NAT, O27.1.1, RING3, RNF3, female sterile homeotic-related gene 1, really interesting new gene 3 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

western blot: 1:10,000-1:40,000

accession no.

NP_005095.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

goat ... BRD2(6046)

Immunogen

The epitope recognized by PLA0066 maps to a region between residues 550 and 600 of human Bromodomain Containing 2 using the numbering given in entry NP_005095.1 (GeneID 6046).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

BRD2 (bromodomain-containing 2) is a nuclear mitogen-activated kinase that plays a role in transcription of cell-cycle-regulated genes by functioning as an adapter protein that recruits E2F and chromatin remodeling factors to E2F response elements.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service