Skip to Content
MilliporeSigma
All Photos(1)

Documents

PLA0031

Sigma-Aldrich

Rabbit anti-TRX2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, gorilla, rhesus monkey, primate, gibbon, chimpanzee

concentration

1 mg/mL

technique(s)

ChIP: 1- 5 μg
immunoprecipitation (IP): 5- 10 μg/mg
western blot: 1:500- 1:5,000

accession no.

NP_055542.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... TRX2(9757)

Immunogen

The epitope recognized by PLA0031 maps to a region between residue 2375 and 2425 of human Trithorax 2 using the numbering given in entry NP_055542.1 (GeneID 9757).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Human trithorax homolog 2 (TRX2) is a member of the MLL sub-family of proteins that are found in Set1-like histone H3 K4 methyltransferase complexes. TRX2 may function as a transactivator that mediates cell adhesion and cytoskeletal events important to cell growth and survival. TRX2 is also known as MLL2. The MLL2 complex includes MLL2 (TRX2), ASH2, RBQ3, WDR5, hDPY-30, and menin. MLL2 has been shown to interact with ER-alpha to regulate estrogen-induced gene expression and has been implicated in tumorigenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service