Skip to Content
MilliporeSigma
All Photos(3)

Documents

PIC0004

Sigma-Aldrich

Protease Inhibitor Cocktail

ReadyShield® ready-to-use solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use in purification of Histidine-tagged proteins, Non-freezing solution

Synonym(s):

Protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.28

product name

ReadyShield® Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, Non-freezing solution

description

Purification of Histidine-Tagged Proteins

form

ready-to-use solution

quality

200

packaging

pkg of 1 mL (in amber polypropylene vial)
pkg of 5 mL (in amber polypropylene vial)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This particular protease inhibitor cocktail omits chelators (e.g., EDTA), to optimize its use for purification of histidine-tagged proteins.1 It contains inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases. The cocktail has been tested on extracts from E. coli and insect cells This cocktail is supplied as a ready-to-use solution using a proprietary non-freezing formulation. ReadyShield PIC0004 is a non-freezing formulation that contains the same inhibitors as is the DMSO based cocktail P8849.

Features and Benefits

  • Does not freeze in storage for quick, convenient use.
  • Tailored for use with Histidine fusion tagged proteins.
  • Suggested as 100 X solution.
  • Reclosable screw-top vials.
  • Can be stored in cooler for approx. 1 month or room temperature for a week.

Preparation Note

The recommended dilution of the cocktail in the biological extract is 1 ml of the cocktail per 100 ml of lysate from 20 g (wet weight) of E. coli cells or 10 g (wet weight) of baculovirus-infected insect (Spodoptera frugiperda) cells.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

ReadyShield is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service