Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PENB

Sigma-Aldrich

Penicillin G benzathine salt hydrate

~1200 U/mg

Synonym(s):

Benzylpenicillin benzathine salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N4O8S2 · C16H20N2 · xH2O
CAS Number:
Molecular Weight:
909.12 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
51101500

specific activity

~1200 U/mg

SMILES string

O.C(CNCc1ccccc1)NCc2ccccc2.CC3(C)S[C@@H]4[C@H](NC(=O)Cc5ccccc5)C(=O)N4[C@H]3C(O)=O.CC6(C)S[C@@H]7[C@H](NC(=O)Cc8ccccc8)C(=O)N7[C@H]6C(O)=O

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A crusted plaque on the right nipple.
Hsien-Yi Chiu et al.
JAMA, 308(4), 403-404 (2012-07-24)
Syphilis with a lichen planus-like pattern (hypertrophic syphilis).
Gianluca Nazzaro et al.
Journal of cutaneous pathology, 39(8), 805-807 (2012-07-04)
Kathryn Margaret Dupnik et al.
Leprosy review, 83(4), 389-393 (2013-04-26)
There are diverse skin manifestations of both leprosy and syphilis. These diseases can appear similar to many other dermatologic conditions as well as to systemic diseases with dermatologic signs. Nodular syphilis is an uncommon type of secondary syphilis. We present
Piasecka Katarzyna et al.
Klinika oczna, 114(1), 50-52 (2012-07-13)
Syphilis is a sexually transmitted, chronic, systemic infection of increasing prevalence. Ocular symptoms may develop at any stage of the disease process. This is to present a case of acute severe bilateral visual loss due to neuroretinitis with papillar oedema
Azithromycin versus penicillin G benzathine for early syphilis.
Xiang-Sheng Chen
Sexually transmitted diseases, 39(11), 900-901 (2012-10-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service