Skip to Content
MilliporeSigma
All Photos(1)

Documents

P8249

Sigma-Aldrich

Pyoverdines−Gallium (III) complex

from Pseudomonas fluorescens, >90% (HPLC)

Synonym(s):

Pyoverdines, Ga-Pv

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

Pseudomonas fluorescens

Assay

>90% (HPLC)

solubility

H2O: ~10 mg/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gallium-pyoverdines act as growth inhibitors of both P. fluorescens and P. aeruginosa possibly due to gallium interference with bacterial iron metabolism.
Pyoverdines, also called pseuobactins and pyoverdins, are fluorescent siderophores that have high affinity for iron (1032 M-1), and are synthesized by fluorescent pseudomonads under iron-deficient growth conditions. Pyoverdines were shown to prevent iron toxicity produced by iron overload in hepatocyte cultures and effectively scavenges the hydroxyl and peroxyl radicals. Pyoverdines are effective in acquiring iron from transferrin and lactoferrin. Pyoverdines are also involved in the suppression of pythium-induced damping-off of tomato and promotion of growth in some higher plants.

Physical form

Supplied as a mixture containing mainly the succinic acid (MW=1231), 2-hydroxy glutaramide (MW=1260), and succinamide (MW=1230) forms of pyoverdines, bound to gallium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service