Skip to Content
MilliporeSigma
All Photos(2)

Documents

P8077

Sigma-Aldrich

Poly-L-threonine

suitable for cell culture, Mol wt 5,000-15,000

Synonym(s):

L-Threonine homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.26

product name

Poly-L-threonine, mol wt 5,000-15,000

form

solid

mol wt

5,000-15,000

technique(s)

FTIR: suitable
cell culture | mammalian: suitable

storage temp.

−20°C

InChI

1S/C10H10N4O2/c1-7-2-4-8(5-3-7)10-11-13-14(12-10)6-9(15)16/h2-5H,6H2,1H3,(H,15,16)

InChI key

QBJSWVLDHWIJDK-UHFFFAOYSA-N

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Bysell et al.
Langmuir : the ACS journal of surfaces and colloids, 25(1), 522-528 (2008-12-09)
The relative importance of electrostatic and nonelectrostatic interactions in peptide-microgel systems was evaluated by micromanipulator-assisted light microscopy, confocal microscopy, and circular dichroism. For this purpose, the interaction of various homopolypeptides with lightly cross-linked polyelectrolyte gel particles ( approximately 70 microm
Shutoku Matsuyama et al.
Journal of virology, 92(19) (2018-07-20)
Middle East respiratory syndrome coronavirus (MERS-CoV) utilizes host cellular proteases to enter cells. A previous report shows that furin, which is distributed mainly in the Golgi apparatus and cycled to the cell surface and endosomes, proteolytically activates the MERS-CoV spike

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service