Skip to Content
MilliporeSigma
All Photos(1)

Documents

P7665

Sigma-Aldrich

(+)-N6-(2-Phenylisopropyl)adenosine

Synonym(s):

(S)-N6-(1-Methyl-2-phenylethyl)adenosine, (S)-PIA, N6-(D-2-Phenylisopropyl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N5O4
CAS Number:
Molecular Weight:
385.42
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

C[C@@H](Cc1ccccc1)Nc2ncnc3n(cnc23)[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A1 adenosine receptor agonist

Other Notes

Less active isomer

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service