Skip to Content
MilliporeSigma
All Photos(1)

Documents

P7338

Sigma-Aldrich

Protease from HIV-1

recombinant, expressed in E. coli, buffered aqueous solution, suitable for cleaving HIV substrate III (Yields 2 peaks which elute before the substrate peak when analyzed by reversed phase HPLC.)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

form

buffered aqueous solution

suitability

suitable for cleaving HIV substrate III (Yields 2 peaks which elute before the substrate peak when analyzed by reversed phase HPLC.)

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Physical form

Solution in dilute HCl, pH 1.6.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Leuthardt et al.
FEBS letters, 326(1-3), 275-280 (1993-07-12)
The gene coding for the HIV-1 protease was cloned in an Escherichia coli expression vector adding three-histidine codons to the amino and carboxy terminus of the protease sequence. Expression of the protease from this construct led to the accumulation of
J M Louis et al.
Nature structural biology, 6(9), 868-875 (1999-08-31)
In the Gag-Pol polyprotein of HIV-1, the 99-amino acid protease is flanked at its N-terminus by a transframe region (TFR) composed of the transframe octapeptide (TFP) and 48 amino acids of the p6pol, separated by a protease cleavage site. The
Christine L Le Maitre et al.
Arthritis research & therapy, 11(5), R137-R137 (2009-09-17)
Loss of intervertebral disc (IVD) matrix and ultimately disc height as a result of 'degeneration' has been implicated as a major cause of low back pain (LBP). The use of anabolic growth factors as therapies to regenerate IVD matrix, hence
Xiaobin Tang et al.
Journal of immunology (Baltimore, Md. : 1950), 182(9), 5446-5452 (2009-04-22)
ITIM-containing receptors play an essential role in modulating immune responses. Leukocyte-associated inhibitory receptor (LAIR)-1, also known as CD305, is an ITIM-containing inhibitory receptor, expressed by all leukocytes, that binds collagens. In this article, we investigate the effect of a conservative
Rafaela C Sartore et al.
PloS one, 6(6), e20667-e20667 (2011-06-16)
The existence of loss and gain of chromosomes, known as aneuploidy, has been previously described within the central nervous system. During development, at least one-third of neural progenitor cells (NPCs) are aneuploid. Notably, aneuploid NPCs may survive and functionally integrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service