Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7022

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phospho(N-palmitoyl)ethanolamine ammonium salt

~99%

Synonym(s):

L-α-Phosphatidylethanolamine, dipalmitoyl, N-palmitoyl ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C53H104NO9P · NH3
CAS Number:
Molecular Weight:
947.40
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

technique(s)

thin layer chromatography (TLC): suitable

functional group

aldehyde
ketone

shipped in

ambient

storage temp.

−20°C

SMILES string

N.CCCCCCCCCCCCCCCC(=O)NCCOP(O)(=O)OC[C@@H](COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C53H104NO9P.H3N/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-51(55)54-46-47-61-64(58,59)62-49-50(63-53(57)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)48-60-52(56)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2;/h50H,4-49H2,1-3H3,(H,54,55)(H,58,59);1H3/t50-;/m1./s1

InChI key

CREASFJXMVEMCZ-VDKVIGKMSA-N

Biochem/physiol Actions

N-Acyl phosphatidylethanolamine has been reported to accumulate in heart muscle damaged by ischemia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Epps et al.
Biochimica et biophysica acta, 618(3), 420-430 (1980-06-23)
A new phospholipid was detected in the infarcted areas of canine myocardium 24 h after ligation of the left descending branch of the coronary artery. Both the central and peripheral areas of the infarct contained significant amounts of this lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service