Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6915

Sigma-Aldrich

Phosphocreatine disodium salt hydrate

~98% (enzymatic)

Synonym(s):

N-(Imino[phosphonoamino]methyl)-N-methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8N3O5PNa2 · x H2O
CAS Number:
Molecular Weight:
255.08 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~98% (enzymatic)

impurities

<0.02% free creatine

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].CN(CC(O)=O)C(=N)NP([O-])([O-])=O

InChI

1S/C4H10N3O5P.2Na.H2O/c1-7(2-3(8)9)4(5)6-13(10,11)12;;;/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);;;1H2/q;2*+1;/p-2

InChI key

KZVYPOKBYRXZKS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phosphocreatine, a high-energy phosphate reservoir in vertebrate and some invertebrate muscles, provides phosphate for ADP-ATP conversion.

Packaging

This type not always available. To avoid delay, we will substitute P 7936 unless we are specifically asked not to do so.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service