Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6847

Sigma-Aldrich

Monoclonal Anti-Interleukin-2−R-Phycoerythrin antibody produced in mouse

clone 5334, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Interleukin-2, Monoclonal Anti-IL-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5334, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IL2(3558)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-2 (IL-2) also refers as T Cell Growth Factor is a cytokine produced by certain subsets of T lymphocytes. It plays various functions like inducing interferon γ and B cell growth factor secretion as well as affecting the activation and proliferation of NK cells. It also has a vital role in modulating the expression of the IL-2 receptor and can facilitate B cell growth and differentiation. Mouse monoclonal anti-Interleukin-2−R-Phycoerythrin antibody reacts specifically with human IL-2.

Specificity

The conjugates will identify and quantitate cells containing intracellular forms of human IL-2 by flow cytometry.

Immunogen

recombinant human IL-2

Application

Monoclonal anti-Interleukin-2−R-Phycoerythrin antibody can be used in flow cytometry assays for identifying intracytoplasmic cytokines and for enumerating cytokine producing cells present in a mixed population.

Physical form

Solution in buffered saline containing 0.5% bovine serum albumin and 0.1% sodium azide.

Preparation Note

Purified antibody is conjugated to R-phycoerythrin (PE).

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective in vivo growth of T lymphocytes from normal human bone marrows.
Morgan DA
Science, 193(4257), 1007-1008 (1967)
T A Waldmann et al.
The Journal of experimental medicine, 160(5), 1450-1466 (1984-11-01)
Using anti-Tac, a monoclonal anti-interleukin 2 (IL-2) receptor antibody, we have explored the possibility that certain activated B cells display receptors for IL-2. Resting normal B cells and unselected B cell lines established from normal individuals were Tac antigen negative.
K A Smith et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(3), 864-868 (1985-02-01)
The cell surface density of high-affinity membrane receptors for the T-lymphocytotrophic hormone interleukin 2 (IL-2) determines the rate of T-cell-cycle progression. Since 10-fold greater numbers of IL-2 receptor molecules were found by using a radiolabeled monoclonal antibody reactive with IL-2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service