Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6102

Sigma-Aldrich

Anti-Potassium Channel Kir2.1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ATFB9, Anti-HHBIRK1, Anti-HHIRK1, Anti-IRK1, Anti-KIR2.1, Anti-LQT7, Anti-SQT3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB CL
clone:
polyclonal
species reactivity:
rat
citations:
5
technique(s):
immunohistochemistry: suitable
western blot (chemiluminescent): 1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ2(3759)
mouse ... Kcnj2(16518)
rat ... Kcnj2(29712)

General description

Potassium voltage-gated channel subfamily J member 2 (KCNJ2) is an inward-rectifying potassium channel. This tetrameric protein possesses two transmembrane helix domains (M1 and M2), an ion-selective P-loop between M1 and M2 and cytoplasmic amino and carboxy-terminal domains. The gene encoding KCNJ2 is localized on human chromosome 17q24.3.

Immunogen

synthetic peptide corresponding to amino acids 392-410 of human Kir2.1 (with additional N-terminal cysteine). This epitope is identical in rabbit, bovine, pig, and guinea pig and highly homologous in rat, mouse and chicken.

Biochem/physiol Actions

Potassium voltage-gated channel subfamily J member 2 (KCNJ2) has been shown to have a role in the differentiation and proliferation of bone marrow-derived macrophages (BMDM) in mice. It allows the inward movement of K+ into the cell and maintains its balance. Mutations in the KCNJ2 gene have been linked to Andersen-Tawil syndrome.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% bovine serum albumin, and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of Kir2.1 in human monocyte-derived foam cell maturation
Wei Zhang
Journal of Cellular and Molecular Medicine (2016)
Genome-wide association study identifies a susceptibility locus for thyrotoxic periodic paralysis at 17q24.3
Ching-Lung C
Nature Genetics (2012)
Upregulation of the inwardly rectifying potassium channel Kir2.1 (KCNJ2) modulates multidrug resistance of small-cell lung cancer under the regulation of miR-7 and the Ras/MAPK pathway
Huanxin Liu
Molecular Cancer (2015)
Novel mutation in the KCNJ2 gene is associated with a malignant arrhythmic phenotype of Andersen-Tawil syndrome.
Fernlund E
Annals of Noninvasive Electrocardiology : The Official Journal of the International Society for Holter and Noninvasive Electrocardiology, Inc (2013)
Association of KCNJ2 Genetic Variants with Susceptibility to Thyrotoxic Periodic Paralysis in Patients with Graves' Disease.
Park S
Experimental and Clinical Endocrinology & Diabetes (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service