Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6085

Sigma-Aldrich

Pipenzolate bromide

BioXtra

Synonym(s):

Benzilic acid 1-ethylpiperidin-3-yl ester methobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H28NO3Br
CAS Number:
Molecular Weight:
434.37
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

product line

BioXtra

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.002%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

Br.CC[N]1(C)CCCC(C1)OC(=O)C(O)(c2ccccc2)c3ccccc3

InChI

1S/C22H28NO3.BrH/c1-3-23(2)16-10-15-20(17-23)26-21(24)22(25,18-11-6-4-7-12-18)19-13-8-5-9-14-19;/h4-9,11-14,20,25H,3,10,15-17H2,1-2H3;1H

InChI key

PRMDDJWOUVPRIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacologic effect of somatostatin on bile formation in the dog. Enhanced ductular reabsorption as the major mechanism of anticholeresis.
E René et al.
Gastroenterology, 84(1), 120-129 (1983-01-01)
Return to Pakistan of pipenzolate plus phenobarbitone.
K I Tahir
Lancet (London, England), 339(8791), 498-498 (1992-02-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service