Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6045

Sigma-Aldrich

D-Pantothenic acid hemicalcium salt

suitable for plant cell culture

Synonym(s):

(R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
12352200

technique(s)

cell culture | plant: suitable

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

Application

Precursor in the biosynthesis of coenzyme A.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Other Notes

Cell culture media are generally supplemented with D-Pantothenic acid (vitamin B5), an essential vitamin required for the synthesis of coenzyme-A (CoA) and the metabolism of proteins, carbohydrates, and fats.

This product has been qualified for use in plant cell culture. Also see P5155 for D-Pantothenic acid qualified for use in animal, insect and plant cell culture.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service