Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5954

Sigma-Aldrich

L-α-Phosphatidylinositol ammonium salt from Glycine max (soybean)

≥97%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol), PI, PtdIns

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211

biological source

soybean

Assay

≥97%

form

powder

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phospholipid membrane component, precursor of inositol phosphates.

Packaging

Packaged under Argon.

Other Notes

Contains primarily linoleic and palmitic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yelena Ugolev et al.
The Journal of biological chemistry, 281(28), 19204-19219 (2006-05-17)
Activation of the phagocyte NADPH oxidase involves the assembly of a membrane-localized cytochrome b559 with the cytosolic components p47(phox), p67(phox), p40(phox), and the GTPase Rac (1 or 2). In resting phagocytes, Rac is found in the cytosol as a prenylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service