Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5710

Sigma-Aldrich

D-Pantothenic acid hemicalcium salt

BioXtra

Synonym(s):

(R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioXtra

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

color

white to off-white

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.01%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

Application

Precursor in the biosynthesis of coenzyme A.

Biochem/physiol Actions

D-Panthenol is the alcohol analogue and biological precursor of D-pantothenic acid. These analogues are precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid may have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol may be used for studies of skin protection emulsions from UV irradiation and as a humectant.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Habig et al.
Journal of biomolecular screening, 14(6), 679-689 (2009-05-28)
High-throughput screening often identifies not only specific, stoichiometrically binding inhibitors but also undesired compounds that unspecifically interfere with the targeted activity by nonstoichiometrically binding, unfolding, and/or inactivating proteins. In this study, the effect of such unwanted inhibitors on several different
Daisaku Kobayashi et al.
Journal of pharmacological sciences, 115(2), 230-234 (2011-01-25)
It has been reported that pantothenic acid (vitamin B5) and panthenol, an alcohol derivative of pantothenic acid, have beneficial moisturizing effects on the skin. However, few studies have investigated the mechanism of action of pantothenic acid on skin tissues. We
X Hui et al.
International journal of cosmetic science, 29(4), 277-282 (2008-05-21)
The in vitro absorption of panthenol into and through the human nail was examined in this study. Panthenol, the alcohol form of pantothenic acid (vitamin B5), is believed to act as a humectant and improve the flexibility and strength of
C F Zanatta et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(1), 70-75 (2009-09-22)
Considering the belief that natural lipids are safer for topical applications and that carotenoids are able to protect cells against photooxidative damage, we have investigated whether topical creams and lotions, produced with Buriti oil and commercial surfactants, can exert photoprotective
Tonio Wiederholt et al.
Experimental dermatology, 18(11), 969-978 (2009-04-29)
Topical application of pantothenate is widely used in clinical practice for wound healing. Previous studies identified a positive effect of pantothenate on migration and proliferation of cultured fibroblasts. However, these studies were mainly descriptive with no molecular data supporting a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service