Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5307

Sigma-Aldrich

Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala-Lys-Lys

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H103N17O16
CAS Number:
Molecular Weight:
1270.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ≥70%

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C1CCCN1)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(=O)NC(C(C)O)C(=O)NC(CC(C)C)C(=O)NC(CO)C(=O)NC(C(C)C)C(=O)NC(C)C(=O)NC(C)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(O)=O

Substrates

Protein kinase C substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Alexander et al.
The Biochemical journal, 268(2), 303-308 (1990-06-01)
Activation of protein kinase C (PKC) in human T lymphocytes is an immediate consequence of mitogenic signalling via the antigen-receptor complex and CD2 antigen. In order to investigate further the signal-transduction pathways which result in PKC activation, we have established
J J Egan et al.
Analytical biochemistry, 175(2), 552-561 (1988-12-01)
A method was devised for assaying protein kinases that phosphorylate either Kemptide, such as cAMP-dependent protein kinase, or a glycogen synthase peptide, which is an excellent substrate for protein kinase C. Upon sequential processing of reaction mixtures through tandem columns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service