Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4480

Sigma-Aldrich

PSTAIRE peptide

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H132N20O26
Molecular Weight:
1741.98
MDL number:
UNSPSC Code:
12352202

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CDK1(983)

Amino Acid Sequence

Glu-Gly-Val-Pro-Ser-Thr-Ala-Ile-Arg-Glu-Ile-Ser-Leu-Leu-Lys-Glu

Biochem/physiol Actions

Cyclin-dependent protein kinase (cdk) highly conserved sequence. Used as an antibody blocking peptide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kamo et al.
Journal of immunological methods, 156(2), 163-170 (1992-12-08)
Mice and rabbits were injected with various forms of a 16 amino acid synthetic peptide representing PSTAIR, the evolutionarily conserved region of the protein kinase p34cdc2, for polyclonal antisera and hybridoma-monoclonal antibody production. Antisera from mice injected with an unconjugated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service