Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4454

Sigma-Aldrich

pGlu-Arg-Thr-Lys-Arg-7-amido-4-methylcoumarin trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

L-Pyroglutamyl-Arg-Thr-Lys-Arg-7-amido-4-methylcoumarin trifluoroacetate salt, Pyr-Arg-Thr-Lys-Arg-7-amido-4-methylcoumarin trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H57N13O9 · xC2HF3O2
Molecular Weight:
827.93 (free base basis)
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@@H](O)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCC(=O)N1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C37H57N13O9.C2HF3O2/c1-19-17-29(53)59-27-18-21(10-11-22(19)27)45-31(54)24(8-5-15-43-36(39)40)47-32(55)23(7-3-4-14-38)49-35(58)30(20(2)51)50-34(57)25(9-6-16-44-37(41)42)48-33(56)26-12-13-28(52)46-26;3-2(4,5)1(6)7/h10-11,17-18,20,23-26,30,51H,3-9,12-16,38H2,1-2H3,(H,45,54)(H,46,52)(H,47,55)(H,48,56)(H,49,58)(H,50,57)(H4,39,40,43)(H4,41,42,44);(H,6,7)/t20-,23+,24+,25+,26+,30+;/m1./s1

InChI key

BHPUHMHLXQHSPN-HUQKKSBQSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Fluorogenic substrate for furin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service