Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4420

Sigma-Aldrich

PHI-27 from rat

≥95% (HPLC)

Synonym(s):

His-Ala-Asp-Gly-Val-Phe-Thr-Ser-Asp-Tyr-Ser-Arg-Leu-Leu-Gly-Gln-Ile-Ser-Ala-Lys-Lys-Tyr-Leu-Glu-Ser-Leu-Ile-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C136H216N36O41
CAS Number:
Molecular Weight:
3011.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CC(C)C)NC(=O)C(CO)NC(=O)C(CCC(O)=O)NC(=O)C(CC(C)C)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(C)NC(=O)C(CO)NC(=O)C(NC(=O)C(CCC(N)=O)NC(=O)CNC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(CCCNC(N)=N)NC(=O)C(CO)NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(CC(O)=O)NC(=O)C(CO)NC(=O)C(NC(=O)C(Cc3ccccc3)NC(=O)C(NC(=O)CNC(=O)C(CC(O)=O)NC(=O)C(C)NC(=O)C(N)Cc4cnc[nH]4)C(C)C)C(C)O)C(C)CC)C(N)=O

Gene Information

rat ... Gpi(292804)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nishizawa et al.
FEBS letters, 183(1), 55-59 (1985-04-08)
The nucleotide sequence analysis of cloned cDNA for VIP precursor from rat cerebral cortex reveals that the precursor contains both rat VIP and PHI-27. The deduced primary structure of rat VIP is identical with human VIP. The amino acid sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service