Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4403

Sigma-Aldrich

Parathyroid Hormone Fragment 39-68 human

≥97% (HPLC), Synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C139H234N46O46
CAS Number:
Molecular Weight:
3285.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](CO)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]2CCCN2C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CO)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]3CCCN3C(=O)[C@H](C)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]4CCCN4C(=O)[C@H](C)N)C(C)C)C(C)C)C(=O)NCC(O)=O

InChI

1S/C139H234N46O46/c1-66(2)52-85(110(205)157-61-106(202)203)172-127(222)93(63-187)179-115(210)77(28-16-19-45-142)164-116(211)81(36-40-101(192)193)166-122(217)88(55-74-59-152-65-158-74)173-128(223)94(64-188)180-119(214)83(38-42-103(196)197)170-132(227)107(69(7)8)181-124(219)87(54-68(5)6)178-133(228)108(70(9)10)182-125(220)89(56-99(145)190)174-123(218)91(58-105(200)201)176-118(213)82(37-41-102(194)195)165-112(207)76(27-15-18-44-141)162-111(206)75(26-14-17-43-140)163-113(208)78(29-20-46-153-137(146)147)168-130(225)97-34-25-51-185(97)136(231)84(31-22-48-155-139(150)151)171-117(212)80(35-39-98(144)189)167-126(221)92(62-186)161-100(191)60-156-109(204)72(12)159-121(216)90(57-104(198)199)175-114(209)79(30-21-47-154-138(148)149)169-129(224)96-33-24-50-184(96)135(230)73(13)160-120(215)86(53-67(3)4)177-131(226)95-32-23-49-183(95)134(229)71(11)143/h59,65-73,75-97,107-108,186-188H,14-58,60-64,140-143H2,1-13H3,(H2,144,189)(H2,145,190)(H,152,158)(H,156,204)(H,157,205)(H,159,216)(H,160,215)(H,161,191)(H,162,206)(H,163,208)(H,164,211)(H,165,207)(H,166,217)(H,167,221)(H,168,225)(H,169,224)(H,170,227)(H,171,212)(H,172,222)(H,173,223)(H,174,218)(H,175,209)(H,176,213)(H,177,226)(H,178,228)(H,179,210)(H,180,214)(H,181,219)(H,182,220)(H,192,193)(H,194,195)(H,196,197)(H,198,199)(H,200,201)(H,202,203)(H4,146,147,153)(H4,148,149,154)(H4,150,151,155)/t71-,72-,73-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,107-,108-/m0/s1

InChI key

PMNYRRYRKDDSKY-DEGCQRHESA-N

Gene Information

human ... PTH(5741)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D'Amour et al.
Journal of immunoassay, 10(2-3), 191-205 (1989-01-01)
Two antisera, C-52 and C-97, raised against bovine (b)PTH(1-84) in guinea pigs, were evaluated with 125I-[tyr53] human (h)PTH(53-84) as tracer and intact hPTH(1-84) and synthetic hPTH(39-84), representative of large carboxylterminal ("C") fragments found in circulation, as standards. In both assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service