Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P4203

Sigma-Aldrich

2-Arachidonoyl-1-palmitoyl-sn-glycero-3-phosphocholine

lyophilized powder

Synonym(s):

PC, 1-Hexadecanoyl-2-([cis,cis,cis,cis]-5,8,11,14-eicosatetraenoyl)-sn-glycero-3-phosphocholine, L-α-Phosphatidylcholine, β-arachidonoyl-γ-palmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H80NO8P
CAS Number:
Molecular Weight:
782.08
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:

Assay

≥95% (TLC)
≥97% (GC)

Quality Level

form

lyophilized powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)O[C@H](COC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC)COP([O-])(=O)OCC[N+](C)(C)C

InChI

1S/C44H80NO8P/c1-6-8-10-12-14-16-18-20-21-22-23-25-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-24-19-17-15-13-11-9-7-2/h14,16,20-21,23,25,28,30,42H,6-13,15,17-19,22,24,26-27,29,31-41H2,1-5H3/b16-14-,21-20-,25-23-,30-28-/t42-/m1/s1

InChI key

PUDHBQQGURLBGJ-IVFHWKNFSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under Argon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Khaselev et al.
Journal of lipid research, 41(4), 564-572 (2000-04-01)
Plasmenyl phospholipids are a structurally unique class of lipids that contain a vinyl ether substituent at the sn-1 position of the glycerol backbone, imparting unique susceptibility to oxidative reactions that may take place at the cell membrane lipid bilayer. Several
S Paglin et al.
The Journal of biological chemistry, 268(16), 11697-11702 (1993-06-05)
Hormonally regulated and particulate-associated phospholipase A2 (PLA2) was detected in endothelial cells from bovine pulmonary artery. The enzyme was solubilized and subjected to initial characterization. PLA2 activity was determined in subcellular fractions from bradykinin (BK)-stimulated and nonstimulated cells by following
N Leitinger et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(21), 12010-12015 (1999-10-16)
We previously have demonstrated that oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphorylcholine (OxPAPC), a component of minimally modified low density lipoprotein (MM-LDL), activates endothelial cells to bind monocytes. 1-Palmitoyl-2- (5-oxovaleroyl)-sn-glycero-3-phosphorylcholine (POVPC) and 1- palmitoyl-2-glutaroyl-sn-glycero-3-phosphorylcholine (PGPC), which are present in OxPAPC, MM-LDL, and atherosclerotic lesions, were
A E Drobnies et al.
Biochemistry, 38(47), 15606-15614 (1999-11-26)
The enzyme CTP:phosphocholine cytidylyltransferase (CT) binds reversibly to membranes and is active only in its membrane-bound form. Membrane lipid composition influences the equilibrium between its soluble and membrane-bound forms. Whereas the enzyme is not activated by phosphatidylcholine (PC) vesicles, it
N Leitinger et al.
Advances in experimental medicine and biology, 433, 379-382 (1997-01-01)
Our group has previously demonstrated that oxidized phospholipids derived from mildly oxidized low density lipoprotein (MM-LDL) or oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphorylcholine (oxPAPC) increase monocyte-endothelial interactions. There are indications that the effects of these phospholipids are receptor mediated. Oxidized phospholipids have previously been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service