Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3982

Sigma-Aldrich

[Pro34]-Peptide YY

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$332.00

$332.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
$332.00

About This Item

Empirical Formula (Hill Notation):
C194H294N54O56
Molecular Weight:
4278.74
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

$332.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PYY(5697)

Amino Acid Sequence

Tyr-Pro-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Pro-Arg-Tyr-NH2

General description

Peptide YY (PYY) is synthesized by L cells in the mucosa of the distal ileum, colon, and rectum. It is mainly expressed by peripheral blood mononuclear cells.[1] Peptide YY is located on human chromosome 17q21.[2]

Biochem/physiol Actions

Peptide YY (PYY) is involved in the modulation of gut motility, food intake and insulin secretion and is associated with obesity and type 2 diabetes.[3] It may possess systemic and paracrine functions.[1] NPY agonist with a preference for the Y1 and Y5 subtypes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TLR ligands and butyrate increase Pyy expression through two distinct but inter-regulated pathways
Larraufie P, et al.
Cellular Microbiology, 19(2), e12648-e12648 (2017)
K Kawakubo et al.
Brain research, 854(1-2), 30-34 (2000-04-28)
We previously reported that intracisternal (i.c.) injection of peptide YY (PYY) and low doses of thyrotropin-releasing hormone (TRH) or TRH analog, RX 77368, increased the resistance of the gastric mucosa to ethanol injury through vagal pathways in rats. The gastroprotective
N D Holliday et al.
Canadian journal of physiology and pharmacology, 78(2), 126-133 (2000-03-29)
This investigation describes the relative potencies of four peptide agonists, namely, peptide YY (PYY), [Leu3l,Pro34]PYY (Pro34pYY), neuropeptide Y (NPY), and [Leu31,Pro34]NPY (Pro34NPY), as antisecretory agents in human, rat, and mouse gastrointestinal preparations. The inhibition of agonist responses by the Y1-receptor
Julia Pia Natascha Holler et al.
Peptides, 58, 78-82 (2014-06-28)
Peptide YY is produced by L cells in the mucosa of the distal ileum, colon, and rectum and may have systemic and paracrine functions. We hypothesized that peptide YY is expressed by peripheral blood mononuclear cells. The purpose of the
Y Hort et al.
Genomics, 26(1), 77-83 (1995-03-01)
Neuropeptide Y (NPY), peptide YY (PYY), and pancreatic polypeptide (PP) are structurally related but functionally diverse peptides, encoded by separate genes and expressed in different tissues. Although the human NPY gene has been mapped to chromosome 7, we demonstrate here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service