Skip to Content
MilliporeSigma
All Photos(1)

Documents

P3650

Sigma-Aldrich

1,2-Dimyristoyl-sn-glycero-3-phosphate monosodium salt

≥99%

Synonym(s):

1,2-Ditetradecanoyl-sn-glycero-3-phosphate monosodium salt, 3-sn-Phosphatidic acid, 1,2-dimyristoyl monosodium salt, L-β,γ-Dimyristoyl-α-phosphatidic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H60NaO8P
CAS Number:
Molecular Weight:
614.77
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

synthetic (organic)

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C31H61O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(32)37-27-29(28-38-40(34,35)36)39-31(33)26-24-22-20-18-16-14-12-10-8-6-4-2;/h29H,3-28H2,1-2H3,(H2,34,35,36);/q;+1/p-1/t29-;/m1./s1

InChI key

UBSPGYHFNIKQIP-XXIQNXCHSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana N Samovich et al.
Chemistry and physics of lipids, 222, 8-14 (2019-04-22)
Effects of quinones and azoles on the formation of steady-state radiolysis products in aqueous solutions of glycerol-1-phosphate and aqueous dispersions of 1,2-dimyristoyl-glycero-3-phosphatidyl-glycerol has been investigated. The data obtained by LC-MS-ESI and spectrophotometric measurements shows that the compounds having quinoid structures
Quoc-Chon Le et al.
Colloids and surfaces. B, Biointerfaces, 123, 150-157 (2014-09-23)
A systematic study was carried out on monolayer films and lipid vesicles to elucidate the interactions between membrane lipids and commercial particles of titanium dioxide TiO2 (TiO2-P25). Pressure-area isotherms of lipids at various pH values were recorded on a Langmuir

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service