Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3545

Sigma-Aldrich

DL-Δ1-Pyrroline-5-carboxylic acid 2,4-dinitrophenylhydrazine hydrochloride double salt

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H14N5O6Cl
CAS Number:
Molecular Weight:
347.71
MDL number:
UNSPSC Code:
12352200

Assay

~95%

storage temp.

2-8°C

SMILES string

Cl.OC(=O)C1CCC=N1.NNc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O

Looking for similar products? Visit Product Comparison Guide

Application

Stable source of glutamic semialdehyde

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties and analysis of a stable derivative of pyrroline-5-carboxylic acid for use in metabolic studies.
V A Mezl et al.
Analytical biochemistry, 74(2), 430-440 (1976-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service