Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2990

Sigma-Aldrich

Prion PrP N-Terminal Peptide Fragment 79-97

solid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

solid

mol wt

27-30 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PRNP(5621)

Amino Acid Sequence

Gly-Gln-Gly-Gly-Gly-Thr-His-Ser-Gln-Trp-Asn-Lys-Pro-Ser-Lys-Pro-Lys-Thr-Asn

Biochem/physiol Actions

N-terminal fragment of the protease resistant prion protein (PrP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service