Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2782

Sigma-Aldrich

1-Palmitoyl-2-(pyren-1-yl)hexanoyl-sn-glycero-3-phosphocholine

Synonym(s):

L-α-Phosphatidylcholine, β-(pyren-1-yl)hexanoyl-γ-palmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H68NO8P
CAS Number:
Molecular Weight:
794.01
MDL number:
UNSPSC Code:
12352211

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@@H](COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCc1ccc2ccc3cccc4ccc1c2c34


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service